siRNA Details
| Gene Symbol: Olfr96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146514.1 | NP_666725.1 | 1 | 396 | ||
| GenBank mRNA | BC120652.1 | - | 1 | 432 | 40129575 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr96| Gene Symbol: Olfr96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146514.1 | NP_666725.1 | 1 | 396 | ||
| GenBank mRNA | BC120652.1 | - | 1 | 432 | 40129575 | |
Set Membership: |
|