siRNA Details
Gene Symbol: Olfr96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146514.1 | NP_666725.1 | 1 | 913 | ||
GenBank mRNA | BC120652.1 | - | 1 | 949 | 40129575 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
