siRNA Details
Gene Symbol: Olfr716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146604.1 | NP_666815.1 | 1 | 919 | ||
GenBank mRNA | BC120510.1 | - | 1 | 1011 | 40129433 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146604.1 | NP_666815.1 | 1 | 919 | ||
GenBank mRNA | BC120510.1 | - | 1 | 1011 | 40129433 |