siRNA Details
| Gene Symbol: Olfr1427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146679.1 | NP_666890.1 | 1 | 811 | ||
| GenBank mRNA | BC127962.1 | - | 1 | 815 | 40134881 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1427| Gene Symbol: Olfr1427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146679.1 | NP_666890.1 | 1 | 811 | ||
| GenBank mRNA | BC127962.1 | - | 1 | 815 | 40134881 | |