siRNA Details
Gene Symbol: Olfr920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146787.2 | NP_666998.2 | 2 | 1368 | ||
GenBank mRNA | AK040176.1 | - | 1 | 515 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146787.2 | NP_666998.2 | 2 | 1368 | ||
GenBank mRNA | AK040176.1 | - | 1 | 515 |