siRNA Details
| Gene Symbol: Olfr1499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146796.1 | NP_667007.1 | 1 | 402 | ||
| GenBank mRNA | BC132138.1 | - | 1 | 472 | 40130415 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1499| Gene Symbol: Olfr1499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146796.1 | NP_667007.1 | 1 | 402 | ||
| GenBank mRNA | BC132138.1 | - | 1 | 472 | 40130415 | |