siRNA Details
Gene Symbol: Olfr1339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146852.2 | NP_667063.1 | 1 | 597 | ||
GenBank mRNA | BC104296.1 | - | 1 | 596 | ||
BC104297.1 | - | 1 | 596 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146852.2 | NP_667063.1 | 1 | 597 | ||
GenBank mRNA | BC104296.1 | - | 1 | 596 | ||
BC104297.1 | - | 1 | 596 |