siRNA Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173781.4 | NP_776142.1 | 6 | 526 | ||
| GenBank mRNA | AB232597.1 | - | 6 | 443 | ||
| AK035893.1 | - | 6 | 485 | |||
| BC059903.1 | - | 2 | 109 | |||
| BC060618.1 | - | 6 | 526 | 6406134 | ||

Rab6b
Genomic Map