siRNA Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173781.4 | NP_776142.1 | 5 | 386 | ||
| GenBank mRNA | AB232597.1 | - | 5 | 303 | ||
| AK035893.1 | - | 5 | 345 | |||
| BC060618.1 | - | 5 | 386 | 6406134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab6b| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173781.4 | NP_776142.1 | 5 | 386 | ||
| GenBank mRNA | AB232597.1 | - | 5 | 303 | ||
| AK035893.1 | - | 5 | 345 | |||
| BC060618.1 | - | 5 | 386 | 6406134 | ||