siRNA Details
| Gene Symbol: B9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013717.2 | NP_038745.1 | 5 | 426 | ||
| GenBank mRNA | AB030483.1 | - | 5 | 426 | ||
| AK003038.1 | - | 5 | 421 | |||
| AK010355.1 | - | 5 | 455 | |||
| AK131701.1 | - | 5 | 417 | |||
| BC008113.1 | - | 5 | 394 | 3483517 | ||

B9d1
Genomic Map