siRNA Details
| Gene Symbol: Dguok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162521.1 | NP_001155993.1 | 5 | 726 | ||
| NM_013764.2 | NP_038792.2 | 5 | 726 | |||
| GenBank mRNA | AJ133749.1 | - | 5 | 651 | ||
| AK035281.1 | - | 5 | 672 | |||
| AK167071.1 | - | 5 | 548 | |||
| AY037862.1 | - | 5 | 656 | |||
| BC094920.1 | - | 5 | 726 | 30055743 | ||
| BQ746075.1 | - | 5 | 640 | |||
| U90524.1 | - | 5 | 646 | |||

Dguok
Genomic Map