siRNA Details
| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081633.1 | NP_001075102.1 | 7 | 951 | ||
| NM_022408.2 | NP_071853.2 | 7 | 954 | |||
| GenBank mRNA | AF037256.1 | - | 7 | 953 | ||
| AK088520.1 | - | 7 | 916 | |||
| AK142684.1 | - | 7 | 919 | |||
| AK148404.1 | - | 7 | 922 | |||
| BC013711.1 | - | 7 | 930 | 3708646 | ||

Dgcr14
Genomic Map