siRNA Details
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013897.2 | NP_038925.1 | 1 | 80 | ||
GenBank mRNA | AA204614.1 | - | 1 | 69 | ||
AF196314.1 | - | 69 | ||||
AK003382.1 | - | 1 | 39 | |||
AK004190.1 | - | 69 | ||||
AK012925.1 | - | 1 | 47 | |||
AK018718.1 | - | 1 | 47 | |||
AK019422.1 | - | 1 | 55 | |||
AK169258.1 | - | 1 | 59 | |||
BC029239.1 | - | 1 | 33 | 4237232 | ||
BY100901.1 | - | 1 | 84 |
