siRNA Details
| Gene Symbol: Klk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174866.2 | NP_777355.1 | 3 | 234 | ||
| GenBank mRNA | BC044756.2 | - | 2 | 209 | ||
| BC128019.1 | - | 3 | 237 | 40136266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk14| Gene Symbol: Klk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174866.2 | NP_777355.1 | 3 | 234 | ||
| GenBank mRNA | BC044756.2 | - | 2 | 209 | ||
| BC128019.1 | - | 3 | 237 | 40136266 | ||