siRNA Details
| Gene Symbol: Dusp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176933.4 | NP_795907.1 | 1, 2 | 567 | ||
| GenBank mRNA | AK053746.1 | - | 1, 2 | 564 | ||
| AK080964.1 | - | 1, 2 | 567 | |||
| AK165991.1 | - | 1, 2 | 883 | |||
| BC118941.1 | - | 1, 2 | 538 | 40092218 | ||
| BC118942.1 | - | 1, 2 | 538 | 40092219 | ||

Dusp4
Genomic Map