siRNA Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081567.1 | NP_001075036.1 | 14 | 6379 | ||
| NM_009158.2 | NP_033184.2 | 14 | 6384 | |||
| GenBank mRNA | AK046409.1 | - | 1 | 184 | ||
| AK048693.1 | - | 1 | 2503 | |||
| AK136881.1 | - | 1 | 2082 | |||

Mapk10
Genomic Map