siRNA Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008997.3 | NP_033023.1 | 5 | 2751 | ||
| GenBank mRNA | AK043096.1 | - | 1 | 117 | ||
| BM935109.1 | - | 1 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11b| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008997.3 | NP_033023.1 | 5 | 2751 | ||
| GenBank mRNA | AK043096.1 | - | 1 | 117 | ||
| BM935109.1 | - | 1 | 230 | |||