siRNA Details
| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049685.1 | - | 1106 | |||
| AK173026.1 | - | 1 | 1354 | |||
| BC147280.1 | - | 1 | 438 | 9056352 | ||
| BC147281.2 | - | 1 | 438 | 9056324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wscd2| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049685.1 | - | 1106 | |||
| AK173026.1 | - | 1 | 1354 | |||
| BC147280.1 | - | 1 | 438 | 9056352 | ||
| BC147281.2 | - | 1 | 438 | 9056324 | ||