siRNA Details
| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177292.3 | NP_796266.2 | 1027 | |||
| GenBank mRNA | AK049685.1 | - | 723 | |||
| AK173026.1 | - | 1 | 971 | |||
| BC096526.1 | - | 1027 | ||||
| BC147280.1 | - | 1 | 55 | 9056352 | ||
| BC147281.2 | - | 1 | 55 | 9056324 | ||

Wscd2
Genomic Map