siRNA Details
| Gene Symbol: Rnf212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089585.1 | - | 2, 3 | 456 | ||
| BC109180.2 | - | 2, 3 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf212| Gene Symbol: Rnf212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089585.1 | - | 2, 3 | 456 | ||
| BC109180.2 | - | 2, 3 | 415 | |||