siRNA Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207261.3 | NP_997144.1 | 3 | 513 | ||
| GenBank mRNA | AK139600.1 | - | 3 | 947 | ||
| AK162136.1 | - | 3 | 516 | |||
| AY325301.1 | - | 3 | 427 | |||
| AY542902.1 | - | 3 | 427 | |||
| BC104132.1 | - | 3 | 427 | |||
| BC104133.1 | - | 3 | 424 | |||
| BC115705.1 | - | 3 | 426 | |||
| BC115887.1 | - | 3 | 420 | |||
| BC127136.1 | - | 3 | 427 | 40045157 | ||
| BC127137.1 | - | 3 | 427 | 40045158 | ||

Kcnk18
Genomic Map