siRNA Details
| Gene Symbol: Wdr72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033500.3 | NP_001028672.2 | 19 | 3611 | ||
| GenBank mRNA | AK139209.1 | - | 2 | 216 | ||
| AK142559.1 | - | 19 | 3591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr72| Gene Symbol: Wdr72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033500.3 | NP_001028672.2 | 19 | 3611 | ||
| GenBank mRNA | AK139209.1 | - | 2 | 216 | ||
| AK142559.1 | - | 19 | 3591 | |||