siRNA Details
| Gene Symbol: Epha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007938.2 | NP_031964.2 | 1 | 130 | ||
| GenBank mRNA | AK090366.1 | - | 1 | 131 | ||
| AK135075.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha6| Gene Symbol: Epha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007938.2 | NP_031964.2 | 1 | 130 | ||
| GenBank mRNA | AK090366.1 | - | 1 | 131 | ||
| AK135075.1 | - | 1 | 114 | |||