siRNA Details
| Gene Symbol: Lrcol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033459.3 | NP_001028631.1 | 1 | 41 | ||
| GenBank mRNA | AK141249.1 | - | 1 | 42 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrcol1| Gene Symbol: Lrcol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033459.3 | NP_001028631.1 | 1 | 41 | ||
| GenBank mRNA | AK141249.1 | - | 1 | 42 | ||