siRNA Details
| Gene Symbol: Hmcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024720.3 | NP_001019891.2 | 1 | 538 | ||
| GenBank mRNA | AK052713.1 | - | 1 | 539 | ||
| BC147549.1 | - | 1 | 406 | |||
| BC147568.1 | - | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmcn1| Gene Symbol: Hmcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024720.3 | NP_001019891.2 | 1 | 538 | ||
| GenBank mRNA | AK052713.1 | - | 1 | 539 | ||
| BC147549.1 | - | 1 | 406 | |||
| BC147568.1 | - | 1 | 406 | |||