siRNA Details
| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027925.1 | - | 1 | 22 | ||
| AK036852.1 | - | 1 | 135 | |||
| AK087735.1 | - | 1 | 133 | |||
| BC076607.1 | - | 1 | 146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330013P04Rik| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027925.1 | - | 1 | 22 | ||
| AK036852.1 | - | 1 | 135 | |||
| AK087735.1 | - | 1 | 133 | |||
| BC076607.1 | - | 1 | 146 | |||