siRNA Details
| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081038.2 | NP_001074507.2 | 14 | 1573 | ||
| GenBank mRNA | BY255452.1 | - | 3 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd16| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081038.2 | NP_001074507.2 | 14 | 1573 | ||
| GenBank mRNA | BY255452.1 | - | 3 | 328 | ||