siRNA Details
| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166282.1 | NP_001159754.1 | 1 | 91 | ||
| GenBank mRNA | AK145264.1 | - | 1 | 93 | ||
| AK166434.1 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc33| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166282.1 | NP_001159754.1 | 1 | 91 | ||
| GenBank mRNA | AK145264.1 | - | 1 | 93 | ||
| AK166434.1 | - | 1 | 76 | |||