siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 8 | 2661 | ||
| GenBank mRNA | AK028824.1 | - | 9 | 2722 | ||
| AK038142.1 | - | 1 | 994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspbap1| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 8 | 2661 | ||
| GenBank mRNA | AK028824.1 | - | 9 | 2722 | ||
| AK038142.1 | - | 1 | 994 | |||