siRNA Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019436.3 | NP_062309.2 | 3 | 348 | ||
| GenBank mRNA | AJ236881.1 | - | 3 | 324 | ||
| AK079882.1 | - | 3 | 350 | |||
| BC064055.1 | - | 3 | 361 | 576098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sit1| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019436.3 | NP_062309.2 | 3 | 348 | ||
| GenBank mRNA | AJ236881.1 | - | 3 | 324 | ||
| AK079882.1 | - | 3 | 350 | |||
| BC064055.1 | - | 3 | 361 | 576098 | ||