siRNA Details
| Gene Symbol: Olfr71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019486.1 | NP_062359.1 | 1 | 18 | ||
| GenBank mRNA | BC100681.1 | - | 1 | 17 | ||
| BC100683.1 | - | 1 | 16 | |||
| BC100684.1 | - | 1 | 17 | |||
| BC100686.1 | - | 1 | 17 | |||
| BC115841.1 | - | 1 | 17 | |||
| BC141882.1 | - | 1 | 18 | 40041946 | ||
| BC141892.1 | - | 1 | 18 | 40041948 | ||

Olfr71
Genomic Map