siRNA Details
| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024414.2 | NP_077725.1 | 6 | 644 | ||
| GenBank mRNA | BC132042.1 | - | 6 | 439 | 40130319 | |
| BC132068.1 | - | 6 | 439 | 40130345 | ||
| D29743.1 | - | 6 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx1b| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024414.2 | NP_077725.1 | 6 | 644 | ||
| GenBank mRNA | BC132042.1 | - | 6 | 439 | 40130319 | |
| BC132068.1 | - | 6 | 439 | 40130345 | ||
| D29743.1 | - | 6 | 471 | |||