siRNA Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291046.1 | NP_001277975.1 | 5 | 690 | ||
| NM_019837.2 | NP_062811.1 | 5 | 682 | |||
| GenBank mRNA | AF264064.1 | - | 5 | 681 | ||
| AK079658.1 | - | 3 | 177 | |||
| BC016534.1 | - | 6 | 588 | 2615904 | ||
| BC046805.1 | - | 5 | 683 | 6829170 | ||
| BU919402.1 | - | 5 | 514 | |||

Nudt3
Genomic Map