siRNA Details
| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 3 | 272 | ||
| GenBank mRNA | AF159050.1 | - | 2 | 171 | ||
| AK002800.1 | - | 3 | 273 | |||
| AK005122.1 | - | 3 | 273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgst1| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 3 | 272 | ||
| GenBank mRNA | AF159050.1 | - | 2 | 171 | ||
| AK002800.1 | - | 3 | 273 | |||
| AK005122.1 | - | 3 | 273 | |||