siRNA Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204239.1 | NP_001191168.1 | 4 | 747 | ||
| NM_001204253.1 | NP_001191182.1 | 3 | 595 | |||
| NM_019985.3 | NP_064369.1 | 5 | 843 | |||
| GenBank mRNA | AF201457.1 | - | 5 | 554 | ||
| AK007925.1 | - | 5 | 823 | |||
| AK154961.1 | - | 98 | ||||
| BC064054.1 | - | 5 | 843 | 574821 | ||
| EF070191.1 | - | 4 | 437 | |||
| EF070192.1 | - | 3 | 282 | |||

Clec1b
Genomic Map