siRNA Details
| Gene Symbol: Olfr1508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020513.2 | NP_065259.1 | 2 | 1474 | ||
| GenBank mRNA | CB174565.1 | - | 1 | 381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1508| Gene Symbol: Olfr1508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020513.2 | NP_065259.1 | 2 | 1474 | ||
| GenBank mRNA | CB174565.1 | - | 1 | 381 | ||