siRNA Details
| Gene Symbol: Lenep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020517.4 | NP_065263.1 | 1 | 164 | ||
| GenBank mRNA | AK053703.1 | - | 177 | |||
| BC006806.1 | - | 307 | 3152957 | |||
| CK627663.1 | - | 1 | 127 | |||
| EL956268.1 | - | 1 | 177 | |||
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC009152.1 | - | 1099 | |||

Lenep
Genomic Map