siRNA Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021028.3 | NP_066356.3 | 7, 8 | 567 | ||
| GenBank mRNA | AF105217.1 | - | 7, 8 | 542 | ||
| AJ249341.2 | - | 7, 8 | 545 | |||
| AK041651.1 | - | 7, 8 | 558 | |||
| AK051286.1 | - | 6, 7 | 487 | |||
| AK087557.1 | - | 7, 8 | 568 | |||
| AK154160.1 | - | 7, 8 | 561 | |||
| AK178795.1 | - | 3, 4 | 192 | |||
| AK180350.1 | - | 2, 3 | 119 | |||
| AK187899.1 | - | 3, 4 | 196 | |||
| AK193655.1 | - | 3, 4 | 192 | |||
| AK196494.1 | - | 3, 4 | 192 | |||
| AK216095.1 | - | 3, 4 | 192 | |||
| BC019982.1 | - | 7, 8 | 559 | 4159742 | ||

Tk2
Genomic Map