siRNA Details
| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022326.3 | NP_071721.2 | 7 | 900 | ||
| GenBank mRNA | AF456463.1 | - | 6 | 809 | ||
| AK005428.1 | - | 7 | 900 | |||
| AK005550.1 | - | 7 | 895 | |||
| AY014777.1 | - | 7 | 865 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsm| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022326.3 | NP_071721.2 | 7 | 900 | ||
| GenBank mRNA | AF456463.1 | - | 6 | 809 | ||
| AK005428.1 | - | 7 | 900 | |||
| AK005550.1 | - | 7 | 895 | |||
| AY014777.1 | - | 7 | 865 | |||