siRNA Details
| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294308.1 | NP_001281237.1 | 2 | 400 | ||
| NM_022414.2 | NP_071859.1 | 2 | 400 | |||
| GenBank mRNA | AJ245945.1 | - | 2 | 197 | ||
| AK140173.1 | - | 2 | 384 | |||
| AK165775.1 | - | 2 | 206 | |||
| BC024263.1 | - | 2 | 400 | 5133442 | ||

Ngb
Genomic Map