siRNA Details
| Gene Symbol: Arl4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025404.3 | NP_079680.1 | 2 | 316 | ||
| GenBank mRNA | AK004126.1 | - | 2 | 297 | ||
| AK013320.1 | - | 2 | 294 | |||
| BC016113.1 | - | 2 | 282 | 4481779 | ||
| BQ830332.1 | - | 2 | 316 | |||

Arl4d
Genomic Map