siRNA Details
| Gene Symbol: Pithd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025411.4 | NP_079687.3 | 5 | 599 | ||
| GenBank mRNA | AF400670.1 | - | 5 | 562 | ||
| AK004208.1 | - | 5 | 600 | |||
| AK013877.1 | - | 3 | 262 | |||
| AK050261.1 | - | 5 | 572 | |||
| BC052695.1 | - | 5 | 499 | 5715038 | ||

Pithd1
Genomic Map