siRNA Details
| Gene Symbol: Rgcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025427.2 | NP_079703.2 | 4, 5 | 533 | ||
| GenBank mRNA | AF276981.1 | - | 4, 5 | 545 | ||
| AK004451.1 | - | 4, 5 | 534 | |||
| AK004705.1 | - | 4, 5 | 529 | |||
| BC049580.1 | - | 4, 5 | 529 | 6741281 | ||
| BC050935.1 | - | 4, 5 | 522 | 6818687 | ||
| CK792184.1 | - | 1, 2 | 405 | |||
| CT010403.1 | - | 4 | 395 | |||

Rgcc
Genomic Map