siRNA Details
| Gene Symbol: Scp2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025490.2 | NP_079766.3 | 1 | 234 | ||
| GenBank mRNA | AK005847.1 | - | 1 | 234 | ||
| BC048653.1 | - | 1 | 234 | 6773017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scp2d1| Gene Symbol: Scp2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025490.2 | NP_079766.3 | 1 | 234 | ||
| GenBank mRNA | AK005847.1 | - | 1 | 234 | ||
| BC048653.1 | - | 1 | 234 | 6773017 | ||