siRNA Details
| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290487.1 | NP_001277416.1 | 4 | 446 | ||
| NM_025504.4 | NP_079780.2 | 4 | 1371 | |||
| GenBank mRNA | AK002370.1 | - | 4 | 426 | ||
| AK009150.1 | - | 4 | 429 | |||
| AK039669.1 | - | 4 | 1372 | |||
| BC024867.1 | - | 3 | 402 | 4951552 | ||

Atp5sl
Genomic Map