siRNA Details
| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290487.1 | NP_001277416.1 | 3 | 269 | ||
| NM_025504.4 | NP_079780.2 | 3 | 1194 | |||
| GenBank mRNA | AK009150.1 | - | 3 | 252 | ||
| AK039669.1 | - | 3 | 1195 | |||
| BC024867.1 | - | 2 | 225 | 4951552 | ||

Atp5sl
Genomic Map