siRNA Details
| Gene Symbol: Ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025509.3 | NP_079785.1 | 2, 3 | 304 | ||
| GenBank mRNA | AK011085.1 | - | 2, 3 | 256 | ||
| AK013011.1 | - | 2, 3 | 263 | |||
| AK135332.1 | - | 2, 3 | 260 | |||
| BC014799.1 | - | 2, 3 | 279 | |||
| BC021935.1 | - | 2, 3 | 278 | 5149713 | ||
| BY228396.1 | - | 2, 3 | 306 | |||

Ostc
Genomic Map