siRNA Details
Gene Symbol: Fam213b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025582.3 | NP_079858.2 | 3 | 323 | ||
GenBank mRNA | AK005188.1 | - | 3 | 324 | ||
AK012999.1 | - | 3 | 322 | |||
BC030453.1 | - | 3 | 317 | 5401948 |
