siRNA Details
| Gene Symbol: Rep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025620.2 | NP_079896.1 | 1 | 307 | ||
| GenBank mRNA | AK008892.1 | - | 1 | 308 | ||
| AK008964.1 | - | 1 | 310 | |||
| BC024344.1 | - | 1 | 258 | 4988964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rep15| Gene Symbol: Rep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025620.2 | NP_079896.1 | 1 | 307 | ||
| GenBank mRNA | AK008892.1 | - | 1 | 308 | ||
| AK008964.1 | - | 1 | 310 | |||
| BC024344.1 | - | 1 | 258 | 4988964 | ||