siRNA Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025697.3 | NP_079973.1 | 3, 4 | 648 | ||
| GenBank mRNA | AK018044.1 | - | 3, 4 | 526 | ||
| AK018153.1 | - | 3, 4 | 649 | |||
| AK018290.1 | - | 3, 4 | 498 | |||
| AK137685.1 | - | 497 | ||||
| AK167306.1 | - | 3, 4 | 490 | |||
| AK169059.1 | - | 3, 4 | 497 | |||
| BC037015.1 | - | 3, 4 | 482 | 5344409 | ||

Spryd7
Genomic Map